Assay Details
| Gene Symbol: SC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009406.1 | NP_001009406.1 | 3 - 4 | 382 | 110 | |
| GenBank mRNA | L46854.1 | - | 3 - 4 | 382 | 110 | |
| X92758.1 | - | 382 | 110 | |||
More Information
Set Membership: |
|

SC5
Genomic Map