Assay Details
| Gene Symbol: CNN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009456.1 | NP_001009456.1 | 5 - 6 | 585 | 61 | |
| XM_027969199.1 | XP_027825000.1 | 5 - 6 | 488 | 61 | ||
| GenBank mRNA | AY169240.1 | - | 4 - 5 | 362 | 61 | |
| AY327118.1 | - | 5 - 6 | 585 | 61 | ||
More Information
Set Membership: |
|

CNN1
Genomic Map