Assay Details
| Gene Symbol: BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040009.2 | NP_001035098.1 | 3 - 4 | 182 | 90 | |
| GenBank mRNA | DQ418490.1 | - | 3 - 4 | 186 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLAP| Gene Symbol: BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040009.2 | NP_001035098.1 | 3 - 4 | 182 | 90 | |
| GenBank mRNA | DQ418490.1 | - | 3 - 4 | 186 | 90 | |