Assay Details
| Gene Symbol: TAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082596.1 | NP_001076065.1 | 1 - 2 | 127 | 97 | |
| XM_012176219.3 | XP_012031609.1 | 2 - 3 | 371 | 97 | ||
| XM_012176220.3 | XP_012031610.1 | 2 - 3 | 365 | 97 | ||
| XM_012176221.3 | XP_012031611.1 | 2 - 3 | 376 | 97 | ||
| XM_012176222.3 | XP_012031612.1 | 2 - 3 | 371 | 97 | ||
| GenBank mRNA | EF207555.1 | - | 1 - 2 | 127 | 97 | |
| EF207556.1 | - | 1 - 2 | 95 | 97 | ||
More Information
Set Membership: |
|

TAC1
Genomic Map