Assay Details
| Gene Symbol: CORT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143891.2 | NP_001137363.2 | 1 - 2 | 185 | 89 | |
| GenBank mRNA | FJ479722.1 | - | 1 - 2 | 186 | 89 | |
More Information
Set Membership: |
|

CORT
Genomic Map