Assay Details
Gene Symbol: ARL5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162556.1 | NP_001156028.1 | 3 - 4 | 254 | 66 | |
XM_015098289.2 | XP_014953775.1 | 4 - 5 | 2456 | 66 | ||
XM_027973939.1 | XP_027829740.1 | 4 - 5 | 2102 | 66 | ||
XM_027973940.1 | XP_027829741.1 | 4 - 5 | 2089 | 66 | ||
XM_027973941.1 | XP_027829742.1 | 2 - 3 | 581 | 66 | ||
GenBank mRNA | FJ969401.1 | - | 3 - 4 | 254 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
