Assay Details
| Gene Symbol: LCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162568.1 | NP_001156040.1 | 5 - 6 | 753 | 72 | |
| XM_012189513.2 | XP_012044903.1 | 5 - 6 | 803 | 72 | ||
| GenBank mRNA | FJ958346.1 | - | 5 - 6 | 753 | 72 | |
More Information
Set Membership: |
|

LCAT
Genomic Map