Assay Details
| Gene Symbol: AKIRIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252176.1 | NP_001239105.1 | 1 - 2 | 240 | 100 | |
| GenBank mRNA | JN390431.1 | - | 1 - 2 | 240 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

AKIRIN2| Gene Symbol: AKIRIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252176.1 | NP_001239105.1 | 1 - 2 | 240 | 100 | |
| GenBank mRNA | JN390431.1 | - | 1 - 2 | 240 | 100 | |