Assay Details
Gene Symbol: PAEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009366.1 | NP_001009366.1 | 1 - 2 | 138 | 62 | |
GenBank mRNA | X04520.1 | - | 138 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
