Assay Details
| Gene Symbol: SFTPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009729.1 | NP_001009729.1 | 3 - 4 | 332 | 56 | |
| GenBank mRNA | AF076634.1 | - | 3 - 4 | 332 | 56 | |
| AF211858.1 | - | 3 - 4 | 311 | 56 | ||
| DQ223566.1 | - | 3 - 4 | 332 | 56 | ||
More Information
Set Membership: |
|

SFTPC
Genomic Map