Assay Details
| Gene Symbol: HEXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126343.1 | NP_001119815.1 | 10 - 11 | 1148 | 64 | |
| XM_027971423.1 | XP_027827224.1 | 11 - 12 | 1248 | 64 | ||
| GenBank mRNA | EU579459.1 | - | 10 - 11 | 1148 | 64 | |
More Information
Set Membership: |
|

HEXA
Genomic Map