Assay Details
| Gene Symbol: MAPK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139455.1 | NP_001132927.1 | 9 - 10 | 814 | 61 | |
| XM_027958332.1 | XP_027814133.1 | 9 - 10 | 806 | 61 | ||
| GenBank mRNA | FJ360444.1 | - | 9 - 10 | 814 | 61 | |
| KF770837.1 | - | 9 - 10 | 814 | 61 | ||
More Information
Set Membership: |
|

MAPK13
Genomic Map