Assay Details
| Gene Symbol: DARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161887.1 | NP_001155359.1 | 6 - 7 | 574 | 102 | |
| GenBank mRNA | FJ969422.1 | - | 6 - 7 | 574 | 102 | |
More Information
Set Membership: |
|

DARS1
Genomic Map