Assay Details
| Gene Symbol: NDUFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185123.1 | NP_001172052.1 | 4 - 5 | 316 | 82 | |
| GenBank mRNA | HM017955.1 | - | 4 - 5 | 316 | 82 | |
More Information
Set Membership: |
|

NDUFB6
Genomic Map