Assay Details
Gene Symbol: HAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195312.1 | NP_001182241.1 | 2 - 3 | 152 | 98 | |
GenBank mRNA | GQ901053.1 | - | 2 - 3 | 152 | 98 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195312.1 | NP_001182241.1 | 2 - 3 | 152 | 98 | |
GenBank mRNA | GQ901053.1 | - | 2 - 3 | 152 | 98 |