Assay Details
| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280714.1 | NP_001267643.1 | 1 - 2 | 225 | 64 | |
| XM_012104920.3 | XP_011960310.1 | 3 - 4 | 517 | 64 | ||
| GenBank mRNA | JX569757.1 | - | 1 - 2 | 227 | 64 | |
More Information
Set Membership: |
|

BMPR1A
Genomic Map