Assay Details
| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308583.1 | NP_001295512.1 | 1 - 2 | 328 | 69 | |
| GenBank mRNA | KC119399.1 | - | 1 - 2 | 328 | 69 | |
More Information
Set Membership: |
|

NPFFR2
Genomic Map