Assay Details
| Gene Symbol: ATP5MC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009396.1 | NP_001009396.1 | 3 - 4 | 172 | 98 | |
| XM_012185111.3 | XP_012040501.1 | 3 - 4 | 203 | 98 | ||
| XM_012185112.3 | XP_012040502.1 | 3 - 4 | 260 | 98 | ||
| GenBank mRNA | X69904.1 | - | 172 | 98 | ||
| Gene Symbol: ATP5MC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974671.1 | XP_027830472.1 | 3 - 4 | 203 | 98 | |
| XM_027974673.1 | XP_027830474.1 | 3 - 4 | 468 | 98 | ||
| XM_027974674.1 | XP_027830475.1 | 3 - 4 | 260 | 98 | ||
More Information
Set Membership: |
|

ATP5MC1
Genomic Map