Assay Details
| Gene Symbol: TSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009747.1 | NP_001009747.1 | 2 - 3 | 319 | 55 | |
| XM_027965743.1 | XP_027821544.1 | 3 - 4 | 440 | 55 | ||
| GenBank mRNA | AF290203.1 | - | 2 - 3 | 319 | 55 | |
More Information
Set Membership: |
|

TSPO
Genomic Map