Assay Details
| Gene Symbol: KRT35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114761.1 | NP_001108233.1 | 4 - 5 | 915 | 108 | |
| XM_027973927.1 | XP_027829728.1 | 5 - 6 | 1818 | 108 | ||
| XM_027973928.1 | XP_027829729.1 | 5 - 6 | 1734 | 108 | ||
| GenBank mRNA | EU216426.1 | - | 4 - 5 | 915 | 108 | |
More Information
Set Membership: |
|

KRT35
Genomic Map