Assay Details
| Gene Symbol: RAB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127283.1 | NP_001120755.1 | 1 - 2 | 76 | 90 | |
| XM_015096239.2 | XP_014951725.1 | 1 - 2 | 297 | 90 | ||
| XM_027970714.1 | XP_027826515.1 | 1 - 2 | 297 | 90 | ||
| GenBank mRNA | EU626203.1 | - | 1 - 2 | 76 | 90 | |
More Information
Set Membership: |
|

RAB28
Genomic Map