Assay Details
| Gene Symbol: MTNR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130938.1 | NP_001124410.1 | 1 - 2 | 738 | 53 | |
| GenBank mRNA | EU679365.1 | - | 1 - 2 | 738 | 53 | |
More Information
Set Membership: |
|

MTNR1B
Genomic Map