Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009214.1 | NP_001009214.1 | 1 - 2 | 101 | 73 | |
| XM_027963883.1 | XP_027819684.1 | 1 - 2 | 615 | 73 | ||
| GenBank mRNA | AF092740.1 | - | 1 - 2 | 101 | 73 | |
More Information
Set Membership: |
|

CTLA4
Genomic Map