Assay Details
| Gene Symbol: AQP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009273.2 | NP_001009273.1 | 3 - 4 | 627 | 104 | |
| GenBank mRNA | AY177613.1 | - | 3 - 4 | 627 | 104 | |
More Information
Set Membership: |
|

AQP5
Genomic Map