Assay Details
| Gene Symbol: RIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126361.1 | NP_001119833.1 | 1 - 2 | 103 | 53 | |
| XM_012174025.2 | XP_012029415.1 | 2 - 3 | 269 | 53 | ||
| XM_012174619.3 | XP_012030009.1 | 2 - 3 | 286 | 53 | ||
| GenBank mRNA | EU596600.1 | - | 1 - 2 | 103 | 53 | |
More Information
Set Membership: |
|

RIT1
Genomic Map