Assay Details
| Gene Symbol: BCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308571.1 | NP_001295500.1 | 1 - 2 | 177 | 103 | |
| GenBank mRNA | FN543098.2 | - | 1 - 2 | 177 | 103 | |
More Information
Set Membership: |
|

BCO1
Genomic Map