Assay Details
| Gene Symbol: PTGFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009789.2 | NP_001009789.1 | 2 - 3 | 899 | 96 | |
| XM_012162160.3 | XP_012017550.1 | 2 - 3 | 1355 | 96 | ||
| XM_012162853.3 | XP_012018243.1 | 2 - 3 | 1402 | 96 | ||
| GenBank mRNA | U73798.1 | - | 2 - 3 | 900 | 96 | |
More Information
Set Membership: |
|

PTGFR
Genomic Map