Assay Details
| Gene Symbol: PTGFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009789.2 | NP_001009789.1 | 3 - 4 | 1050 | 77 | |
| GenBank mRNA | U73798.1 | - | 3 - 4 | 1051 | 77 | |
More Information
Set Membership: |
|

PTGFR
Genomic Map