Assay Details
| Gene Symbol: FCER1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123004.1 | NP_001116476.1 | 4 - 5 | 640 | 114 | |
| GenBank mRNA | Y18205.1 | - | 640 | 114 | ||
More Information
Set Membership: |
|

FCER1A
Genomic Map