Assay Details
| Gene Symbol: DTNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126349.1 | NP_001119821.1 | 3 - 4 | 154 | 71 | |
| GenBank mRNA | EU583472.1 | - | 3 - 4 | 154 | 71 | |
More Information
Set Membership: |
|

DTNBP1
Genomic Map