Assay Details
| Gene Symbol: FBXL21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129738.1 | NP_001123210.1 | 2 - 3 | 487 | 114 | |
| GenBank mRNA | EU239380.1 | - | 2 - 3 | 487 | 114 | |
More Information
Set Membership: |
|

FBXL21P
Genomic Map