Assay Details
| Gene Symbol: SFTPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009728.2 | NP_001009728.2 | 3 - 4 | 412 | 85 | |
| GenBank mRNA | AF076633.1 | - | 3 - 4 | 335 | 85 | |
| AF211856.1 | - | 3 - 4 | 412 | 85 | ||
More Information
Set Membership: |
|

SFTPA1
Genomic Map