Assay Details
| Gene Symbol: RRAGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161867.1 | NP_001155339.1 | 6 - 7 | 1043 | 89 | |
| GenBank mRNA | FJ943975.1 | - | 6 - 7 | 1043 | 89 | |
More Information
Set Membership: |
|

RRAGC
Genomic Map