Assay Details
| Gene Symbol: PIGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280695.1 | NP_001267624.1 | 3 - 4 | 385 | 93 | |
| XM_012159160.2 | XP_012014550.2 | 3 - 4 | 422 | 93 | ||
| XM_027965739.1 | XP_027821540.1 | 3 - 4 | 422 | 93 | ||
| GenBank mRNA | JX534535.1 | - | 3 - 4 | 387 | 93 | |
More Information
Set Membership: |
|

PIGF
Genomic Map