Assay Details
| Gene Symbol: GCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287471.1 | NP_001274400.1 | 5 - 6 | 866 | 56 | |
| GenBank mRNA | KC237373.1 | - | 5 - 6 | 866 | 56 | |
More Information
Set Membership: |
|

GCK
Genomic Map