Assay Details
| Gene Symbol: CTSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308587.1 | NP_001295516.1 | 7 - 8 | 790 | 60 | |
| XM_004004452.3 | XP_004004501.1 | 8 - 9 | 974 | 60 | ||
| XM_012129056.3 | XP_011984446.1 | 8 - 9 | 883 | 60 | ||
| XM_012129063.3 | XP_011984453.1 | 8 - 9 | 910 | 60 | ||
| XM_015092747.2 | XP_014948233.1 | 4 - 5 | 418 | 60 | ||
| XM_027963893.1 | XP_027819694.1 | 8 - 9 | 910 | 60 | ||
| GenBank mRNA | KC699918.1 | - | 7 - 8 | 790 | 60 | |
More Information
Set Membership: |
|

CTSB
Genomic Map