Assay Details
| Gene Symbol: YEATS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163059.1 | NP_001156531.1 | 6 - 7 | 517 | 89 | |
| GenBank mRNA | FJ958351.1 | - | 6 - 7 | 517 | 89 | |
More Information
Set Membership: |
|

YEATS4
Genomic Map