Assay Details
| Gene Symbol: ARNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287465.1 | NP_001274394.1 | 10 - 11 | 956 | 67 | |
| GenBank mRNA | EU340263.1 | - | 1 - 2 | 75 | 67 | |
| HF912413.1 | - | 10 - 11 | 956 | 67 | ||
More Information
Set Membership: |
|

ARNT
Genomic Map