Assay Details
| Gene Symbol: CR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009724.1 | NP_001009724.1 | 12 - 13 | 2696 | 67 | |
| GenBank mRNA | AF038131.1 | - | 12 - 13 | 2696 | 67 | |
More Information
Set Membership: |
|

CR2
Genomic Map