Assay Details
| Gene Symbol: PRLHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126347.1 | NP_001119819.1 | 1 | 319 | 91 | |
| GenBank mRNA | EU194571.1 | - | 1 | 319 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRLHR| Gene Symbol: PRLHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126347.1 | NP_001119819.1 | 1 | 319 | 91 | |
| GenBank mRNA | EU194571.1 | - | 1 | 319 | 91 | |
Set Membership: |
|