Assay Details
| Gene Symbol: AKIRIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127672.1 | NP_001121144.1 | 2 - 3 | 377 | 80 | |
| GenBank mRNA | EU650274.1 | - | 2 - 3 | 377 | 80 | |
More Information
Set Membership: |
|

AKIRIN1
Genomic Map