Assay Details
| Gene Symbol: SLC39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163600.1 | NP_001157072.1 | 2 - 3 | 318 | 84 | |
| XM_012104332.3 | XP_011959722.1 | 3 - 4 | 642 | 84 | ||
| XM_012104856.2 | XP_011960246.1 | 4 - 5 | 516 | 84 | ||
| XM_012105400.3 | XP_011960790.1 | 3 - 4 | 457 | 84 | ||
| GenBank mRNA | FJ937953.1 | - | 2 - 3 | 318 | 84 | |
More Information
Set Membership: |
|

SLC39A1
Genomic Map