Assay Details
| Gene Symbol: ICAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009731.1 | NP_001009731.1 | 4 - 5 | 944 | 119 | |
| XM_027969187.1 | XP_027824988.1 | 4 - 5 | 975 | 119 | ||
| XM_027969188.1 | XP_027824989.1 | 3 - 4 | 1797 | 119 | ||
| GenBank mRNA | AF110984.1 | - | 4 - 5 | 944 | 119 | |
More Information
Set Membership: |
|

ICAM1
Genomic Map