Assay Details
| Gene Symbol: PSPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126360.1 | NP_001119832.1 | 4 - 5 | 568 | 70 | |
| XM_012103911.3 | XP_011959301.1 | 5 - 6 | 840 | 70 | ||
| XM_027961103.1 | XP_027816904.1 | 5 - 6 | 747 | 70 | ||
| GenBank mRNA | EU583474.1 | - | 4 - 5 | 568 | 70 | |
More Information
Set Membership: |
|

PSPH
Genomic Map