Assay Details
| Gene Symbol: IGFBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134302.1 | NP_001127774.1 | 3 - 4 | 641 | 118 | |
| GenBank mRNA | EU797606.1 | - | 3 - 4 | 641 | 118 | |
| EU882037.1 | - | 3 - 4 | 641 | 118 | ||
| S77394.1 | - | 3 - 4 | 578 | 118 | ||
More Information
Set Membership: |
|

IGFBP4
Genomic Map