Assay Details
| Gene Symbol: ASMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306120.1 | NP_001293049.1 | 661 | 77 | ||
| GenBank mRNA | JQ647512.1 | - | 584 | 77 | ||
| KC290950.1 | - | 661 | 77 | |||
More Information
Set Membership: |
|

ASMT
Genomic Map