Assay Details
| Gene Symbol: CA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009408.2 | NP_001009408.1 | 6 - 7 | 869 | 72 | |
| GenBank mRNA | Y07785.1 | - | 870 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009408.2 | NP_001009408.1 | 6 - 7 | 869 | 72 | |
| GenBank mRNA | Y07785.1 | - | 870 | 72 | ||