Assay Details
| Gene Symbol: SLC2A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009770.1 | NP_001009770.1 | 7 - 8 | 1079 | 67 | |
| GenBank mRNA | L39214.1 | - | 7 - 8 | 1079 | 67 | |
More Information
Set Membership: |
|

SLC2A3
Genomic Map