Assay Details
| Gene Symbol: PSMB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131030.1 | NP_001124502.1 | 3 - 4 | 405 | 73 | |
| GenBank mRNA | EU797604.1 | - | 3 - 4 | 405 | 73 | |
More Information
Set Membership: |
|

PSMB8
Genomic Map