Assay Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145180.1 | NP_001138652.1 | 1 - 2 | 146 | 82 | |
| GenBank mRNA | DQ223551.1 | - | 1 - 2 | 132 | 82 | |
| FJ546079.1 | - | 1 - 2 | 146 | 82 | ||
More Information
Set Membership: |
|

FABP5
Genomic Map